Check Order Status
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
10 kg
100 g
500 g
1 kg
3 kg
Packaging:
View all
Poly Pail
Catalog number
FCMS318-10
Price (MYR)
2,133.00
Add to cart
Quantity:
10 kg
Packaging:
Poly Pail
Request bulk or custom format
Price (MYR)
2,133.00
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide
pH
14
Cleanup Symbol
Caustic
Color
White
Boiling Point
1390°C
View more