Check Order Status
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
100 g
500 g
1 kg
3 kg
10 kg
Packaging:
View all
Poly Bottle
Catalog number
FCMS318-100
Price (MYR)
446.00
Add to cart
Quantity:
100 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
446.00
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide
pH
14
Cleanup Symbol
Caustic
Color
White
Boiling Point
1390°C
View more