Check Order Status
1,2,4-Trichlorobenzene (HPLC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
1,2,4-Trichlorobenzene (HPLC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMO4846-4
4 L
Amber Glass, EcoSafPak™
Catalog number
FCMO4846-4
Price (MYR)
1,865.00
Add to cart
Quantity:
4 L
Packaging:
Amber Glass, EcoSafPak™
Request bulk or custom format
Price (MYR)
1,865.00
Add to cart
Specifications
Chemical Name or Material
1, 2, 4-Trichlorobenzene
Melting Point
16°C
CAS
120-82-1
ChemAlert Storage Symbol
Blue
Density
1.450g/cm³
View more