Check Order Status
Marble Chips, Fisher Chemical
Element Search
Structure Search
Marble Chips, Fisher Chemical
For Kipp generator
Have Questions?
Catalog Number
Quantity
Packaging
FCMM123-3
3 kg
Poly Bottle
Catalog number
FCMM123-3
Price (MYR)
-
Quantity:
3 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Marble Chips
Melting Point
897.8°C
CAS
1317-65-3
ChemAlert Storage Symbol
Gray
Color
Gray
View more