Check Order Status
n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis
Have Questions?
Change view
Quantity:
4 L
Packaging:
Catalog number
FCMH306-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS
122-99-6
IUPAC Name
2-phenoxyethan-1-ol
Molecular Formula
C8H10O2
InChI Key
QCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILES
OCCOC1=CC=CC=C1
View more
Specifications
Specification Sheet
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.5360 to 1.5400 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Tested for GC/MS suitability
Also meets ACS specifications
Intended for research use only