Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Fisher Chemical
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, 500mL, 57 deg.C, Colorless, 36.46g/mol, -35 deg.C, 11324, Liquid, 1.27, 125mbar at 20 deg.C, 1.8 mPaS at 15 deg.C
Have Questions?
Catalog NumberQuantity
FCMA508-P500500 mL
Catalog number FCMA508-P500
Price (MYR)
743.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
743.00
Add to cart
Specifications
pH1
ColorColorless
CAS7647-01-0
Melting Point-35°C
Boiling Point57°C
View more
ClH, CAS Number-7647-01-0, 500mL, 57 deg.C, Colorless, 36.46g/mol, -35 deg.C, 11324, Liquid, 1.27, 125mbar at 20 deg.C, 1.8 mPaS at 15 deg.C