Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Fisher Chemical
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
FEP Bottle
Catalog number FCMA467-500
Price (MYR)
3,704.00
Add to cart
Quantity:
500 mL
Packaging:
FEP Bottle
Request bulk or custom format
Price (MYR)
3,704.00
Add to cart
Chemical Identifiers
CAS7440-38-2
IUPAC Namearsenic
Molecular FormulaAs
InChI KeyRQNWIZPPADIBDY-UHFFFAOYSA-N
SMILES[As]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -20 mesh
Total Metal Impurities≤1.0%
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.