Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Fisher Chemical
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FCMA461-4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS80-18-2
IUPAC Namemethyl benzenesulfonate
Molecular FormulaC7H8O3S
InChI KeyCZXGXYBOQYQXQD-UHFFFAOYSA-N
SMILESCOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Refractive index1.5150 to 1.5180 (20°C, 589 nm)
Appearance (Form)Liquid
HPLC>=98.5 %
Infrared spectrumConforms
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm