Check Order Status
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Suitable as a mobile phase additive for LC/MS and UHPLC/MS
Have Questions?
Change view
Quantity:
10 x 1 mL
50 mL
Packaging:
View all
Ampules
Catalog number
FCMA117-10X1AMP
Price (MYR)
1,338.00
Add to cart
Quantity:
10 x 1 mL
Packaging:
Ampules
Request bulk or custom format
Price (MYR)
1,338.00
Add to cart
Specifications
Absorbance
0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
Chemical Name or Material
Formic Acid
Cleanup Symbol
Acid
pH
2.1
CAS
64-18-6
View more