Check Order Status
D-(+)-Maltose Monohydrate, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
D-(+)-Maltose Monohydrate, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP684-500
500 g
Poly Bottle
Catalog number
FBRBP684-500
Price (MYR)
765.00
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
765.00
Add to cart
Chemical Identifiers
CAS
4238-71-5
IUPAC Name
1-benzyl-1H-imidazole
Molecular Formula
C10H10N2
InChI Key
KKKDZZRICRFGSD-UHFFFAOYSA-N
SMILES
C(N1C=CN=C1)C1=CC=CC=C1
View more
Specifications
Specification Sheet
Appearance (Form)
Crystalline powder or crystals
Melting point
68°C to 72°C
Infrared spectrum
Conforms
Appearance (Color)
White to brown
Titration with HClO4
98.5 to 101.5 %
Maltose is suitable for use in cell culture systems requiring sugar additives.