EZ-Run™ Prestained Rec Protein Ladder, Fisher BioReagents
EZ-Run&trade; Prestained <i>Rec</i> Protein Ladder, Fisher BioReagents
Actual product may vary
EZ-Run&trade; Prestained <i>Rec</i> Protein Ladder, Fisher BioReagents
EZ-Run&trade; Prestained <i>Rec</i> Protein Ladder, Fisher BioReagents
Fisher Chemical
EZ-Run™ Prestained Rec Protein Ladder, Fisher BioReagents
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySufficient For
FBRBP3603-12 x 500 μL, 200 Loadings200 Loadings
FBRBP3603-500500 μL, 100 Loadings100 Loadings
Catalog number FBRBP3603-1
Price (MYR)
2,043.00
Add to cart
Quantity:
2 x 500 μL, 200 Loadings
Sufficient For:
200 Loadings
Price (MYR)
2,043.00
Add to cart
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
  • Ladder is ideal for monitoring protein separation during SDS-PAGE, verifying Western blot transfer efficiency and approximating the molecular weight of blotted proteins
  • Supplied in gel loading buffer
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.
This product is manufactured under the license for Strep-tag™ technology covered by U.S. Patent No. 5,506,121; U.K. Patent No. 2272698; and French Patent No. 93 13 066.
Specifications
ColorBlue
Product TypeProtein Molecular Weight Marker/Ladder
FormLiquid
FormulationSupplied in gel-loading buffer
For Use With (Application)Monitoring protein separation during SDS-PAGE
Quantity2 x 500 μL, 200 Loadings
Range10-170 kDa
Storage Requirements−20°C
Sufficient For200 Loadings
Unit Size