Check Order Status
Sodium Hydroxide (White Pellets), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (White Pellets), Fisher BioReagents™
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change view
Quantity:
2.5 kg
500 g
Packaging:
Poly Bottle
Catalog number
FBRBP359-212
Price (MYR)
807.00
Add to cart
Quantity:
2.5 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
807.00
Add to cart
Chemical Identifiers
CAS
300-08-3
IUPAC Name
hydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
Molecular Formula
C8H14BrNO2
InChI Key
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
SMILES
[H+].[Br-].COC(=O)C1=CCCN(C)C1
View more
Specifications
Specification Sheet
Titration Argentometric
>=98.0 %
Appearance (Color)
White to light yellow
Melting point
168°C to 174°C
Infrared spectrum
Conforms
Appearance (Form)
Powder
Sodium Hydroxide (White Pellets)
Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications.