Sodium Pyruvate (White Powder), Fisher BioReagents
Sodium Pyruvate (White Powder), Fisher BioReagents
Actual product may vary
Sodium Pyruvate (White Powder), Fisher BioReagents
Sodium Pyruvate (White Powder), Fisher BioReagents
Fisher Chemical
Sodium Pyruvate (White Powder), Fisher BioReagents
CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP356-100100 gGlass Bottle
Catalog number FBRBP356-100
Price (MYR)
455.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Price (MYR)
455.00
Add to cart
Glass Bottle; 100g
This sodium salt is used in buffers for molecular biology and cell culture applications.
Specifications
Chemical Name or MaterialSodium Pyruvate
CAS113-24-6
ChemAlert Storage SymbolGray
Melting Point300°C
ColorWhite
Recommended StorageRT
pH5.5 to 6.5
Assay Percent Range≥99 %
PackagingGlass Bottle
Molecular FormulaC3H3NaO3
Linear FormulaCH3COCO2Na
Quantity100 g
Synonymsodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
InChI KeyDAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILESCC(=O)C(=O)[O-].[Na+]
IUPAC Namesodium;2-oxopropanoate
Molecular Weight (g/mol)110.044
PubChem CID23662274
ChEBICHEBI:50144
Formula Weight110.04
Percent Purity≥99%
Physical FormPowder or Solid
Unit Size