Check Order Status
L-Ascorbic Acid (White Crystalline Powder), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
L-Ascorbic Acid (White Crystalline Powder), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP351-500
500 g
Glass Bottle
Catalog number
FBRBP351-500
Price (MYR)
735.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
735.00
Add to cart
Specifications
Alpha Vector
Ascorbic Acid
Chemical Name or Material
L-Ascorbic Acid
CAS
50-81-7
ChemAlert Storage Symbol
Gray
Melting Point
190°C
View more
Ascorbic Acid is suitable for use in tissue culture systems requiring additives.