Check Order Status
Imidazole (Molecular Biology), Fisher BioReagents™,50g
Element Search
Structure Search
Fisher Chemical
Imidazole (Molecular Biology), Fisher BioReagents™,50g
Commonly used buffering agent
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP305-50
50 g
Poly Bottle
Catalog number
FBRBP305-50
Price (MYR)
732.00
Add to cart
Quantity:
50 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
732.00
Add to cart
Specifications
Chemical Name or Material
Imidazole
CAS
288-32-4
ChemAlert Storage Symbol
White
Absorbance
0.01 max. (0.1M solution) at 260nm
Health Hazard 2
Signal Word: DANGER!
View more
Imidazole is an excellent biological buffer with a usable pH range of 6.2 to 7.8.