Check Order Status
MES (Fine White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
MES (Fine White Crystals), Fisher BioReagents
Commonly used buffering agent
Have Questions?
Catalog Number
Quantity
FBRBP300-100
100 g
Catalog number
FBRBP300-100
Price (MYR)
460.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
460.00
Add to cart
Specifications
Chemical Name or Material
MES
CAS
145224-94-8
ChemAlert Storage Symbol
Gray
Melting Point
300°C
Color
White
View more
This biological buffer has a usable pH range of 5.5 to 6.7.