Check Order Status
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
For nucleic acid precipitation
Have Questions?
Change view
Quantity:
500 mL
1 L
2.5 L
4 L
Catalog number
FBRBP2618500
Price (MYR)
380.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS
104-98-3
IUPAC Name
(2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
Molecular Formula
C6H6N2O2
InChI Key
LOIYMIARKYCTBW-OWOJBTEDSA-N
SMILES
OC(=O)C=CC1=CN=CN1
View more
Specifications
Specification Sheet
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
217°C to 220°C
HPLC
>=97.5 %
View more
For nucleic acid precipitation
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.