exACTGene™ DNA Ladders
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
Actual product may vary
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
exACTGene™ DNA Ladders
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of BandsFor Use With (Application)
FBRBP2573-10010General purpose, small size DNA
FBRBP2578-10013General purpose, large digested DNA
FBRBP2579-10019General purpose, wide size range
Catalog number FBRBP2573-100
Price (MYR)
304.63
Add to cart
No. of Bands:
10
For Use With (Application):
General purpose, small size DNA
Price (MYR)
304.63
Add to cart
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes
Specifications
ColorColorless
Product TypeDNA Molecular Weight Marker/Ladder
FormLiquid
FormulationPremixed with loading dye
For Use With (Application)General purpose, small size DNA
No. of Bands10
Quantity100 Lanes
Storage RequirementsStable at room temperature for two years.
Sufficient For100 Lanes
Unit Size