Mitomycin C, Fisher BioReagents™
Mitomycin C, Fisher BioReagents™
Mitomycin C, Fisher BioReagents™
Mitomycin C, Fisher BioReagents™
Fisher Chemical
Mitomycin C, Fisher BioReagents™
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 mg
10 mg
20 mg
Packaging:
Amber Glass
Catalog number FBRBP2531-2
Price (MYR)
926.00
Add to cart
Quantity:
2 mg
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
926.00
Add to cart
Specifications
Chemical Name or MaterialMitomycin C
CAS50-07-7
ChemAlert Storage SymbolBlue
Health Hazard 2WARNING!
Health Hazard 3Emergency Overview
Toxic if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is not required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:0
View more
Mitomycin C
Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.
Recommended Storage: 4°C. Keep containers tightly closed in a dry, cool and well-ventilated place. Keep refrigerated.