Magnesium Sulfate Heptahydrate, Fisher BioReagents
Magnesium Sulfate Heptahydrate, Fisher BioReagents
Actual product may vary
Magnesium Sulfate Heptahydrate, Fisher BioReagents
Magnesium Sulfate Heptahydrate, Fisher BioReagents
Fisher Chemical
Magnesium Sulfate Heptahydrate, Fisher BioReagents
CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP213-11 kgPoly Bottle
Catalog number FBRBP213-1
Price (MYR)
503.00
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Price (MYR)
503.00
Add to cart
Poly Bottle; 1kg
Magnesium Sulfate is suitable for use in the preparation of buffer solutions.
Specifications
Chemical Name or MaterialMagnesium Sulfate Heptahydrate
Melting Point770°C
CAS10034-99-8
ChemAlert Storage SymbolGray
ColorWhite
pH5 to 8.2
Recommended StorageRT
Physical FormSolid
Quantity1 kg
Assay Percent Range98.0 to 102.0 %
Molecular FormulaH14MgO11S
MDL NumberMFCD00149785
Merck Index15, 5755
Synonymmagnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Namemagnesium;sulfate;heptahydrate
Molecular Weight (g/mol)246.47
PubChem CID24843
ChEBICHEBI:31795
Formula Weight246.47
Percent Purity98.0 to 102.0%
IdentificationPass Test
PackagingPoly Bottle
Vapor Pressure<0.1 mmHg at 20°C
Density1.678g/cm³
Unit Size