Check Order Status
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP179-100
100 g
Glass Bottle
Catalog number
FBRBP179-100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ammonium Persulfate
Melting Point
120°C
CAS
7727-54-0
ChemAlert Storage Symbol
Yellow
Color
White
View more
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis)
Electrophoresis-grade Ammonium Persulfate is an initiator of acrylamide polymerization for the preparation of polyacrylamide electrophoresis gels.