Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
Actual product may vary
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
Fisher Chemical
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
For isolation of RNA from cell extracts
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FBRBP178-11 kg
FBRBP178-500500 g
Catalog number FBRBP178-1
Price (MYR)
1,082.00
Add to cart
Quantity:
1 kg
Price (MYR)
1,082.00
Add to cart
For isolation of RNA from cell extracts
Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.
Specifications
Chemical Name or MaterialGuanidine Hydrochloride
CAS50-01-1
Assay Percent Range≥99.5%
Synonymguanidine hydrochloride
Recommended StorageStore at Room Temperature
Quantity1 kg
Merck Index15, 4597
Melting Point178°C
Molecular FormulaCH6ClN3
Formula Weight95.53
Physical FormSolid
ColorWhite
Purity≥99.5%
Unit Size