Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Fisher Chemical
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
View all
Poly Bottle
Catalog number FBRBP168-1
Price (MYR)
444.00
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Specifications
Chemical Name or MaterialBoric Acid
pH3.8 to 4.8
ColorWhite
CAS10043-35-3
ChemAlert Storage SymbolGray
View more
This high-purity electrophoresis-grade reagent is used in the preparation of Tris/Borate/EDTA electrophoresis buffers.