Check Order Status
Agarose (Medium-EEO/Protein Electrophoresis Grade), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Agarose (Medium-EEO/Protein Electrophoresis Grade), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP161-100
100 g
Catalog number
FBRBP161-100
Price (MYR)
1,043.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,043.00
Add to cart
Specifications
Chemical Name or Material
Agarose
Name Note
Medium-EEO
Physical Form
Powder/Solid
CAS
9012-36-6
ChemAlert Storage Symbol
Gray
View more
Ideal alternative to polyacrylamide for serum protein electrophoresis. Immunoelectrophoresis. Electrophoresis of nucleic acids.