Check Order Status
Acrylamide: Bis-Acrylamide 19:1 (40% Solution/Electrophoresis), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Acrylamide: Bis-Acrylamide 19:1 (40% Solution/Electrophoresis), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP1406-1
1 L
Catalog number
FBRBP1406-1
Price (MYR)
791.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
791.00
Add to cart
Specifications
Chemical Name or Material
Acrylamide: Bis-Acrylamide 19:1
Name Note
40% Solution
Physical Form
Liquid
CAS
79-06-1
ChemAlert Storage Symbol
Blue
View more
This Acrylamide Solution is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.