Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
Actual product may vary
Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
For DNA sequencing and other biological applications such as DNA/RNA electrophoresis | CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1185-500500 mLGlass
Catalog number FBRBP1185-500
Price (MYR)
530.00
Add to cart
Quantity:
500 mL
Packaging:
Glass
Price (MYR)
530.00
Add to cart
For DNA sequencing and other biological applications such as DNA/RNA electrophoresis
Aldehyde-free Acetic Acid is suitable for DNA sequencing and other biological applications.
Specifications
Chemical Name or MaterialAcetic Acid, Glacial
Cleanup SymbolAcid
Name NoteAldehyde-free
pH2.5
CAS64-19-7
ChemAlert Storage SymbolRed
Melting Point16°C
Boiling Point117°C
ColorColorless
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:2
Instability:0
Recommended StorageRT
Assay Percent Range≥99.7 %
Molecular FormulaC2H4O2
Linear FormulaCH3COOH
MDL NumberMFCD00036152
UN Number2789
Merck Index15, 54
Synonymethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
IUPAC Nameacetic acid
Molecular Weight (g/mol)60.05
PubChem CID176
ChEBICHEBI:15366
Formula Weight60.05
Percent Purity≥99.7%
GradeSequencing
Density1.048g/cm³
Quantity500 mL
PackagingGlass
Physical FormLiquid
Residue after Evaporation8ppm max.
Vapor Pressure1.52 kPa at 20°C
Unit Size