Bottle Top Dispenser with Re-Circulation Valve
Bottle Top Dispenser with Re-Circulation Valve
Actual product may vary
Fisherbrand
Bottle Top Dispenser with Re-Circulation Valve
Provides a contemporary design coupled with a recirculation valve. Bottle Top Dispenser with Re-Circulation Valve has enhanced chemical compatibility and smooth piston movement.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FBD1368152910 to 100 mL
FIS13-681-5250.5 to 5 mL
FBD136815261 to 10 mL
FIS13-681-5261 to 10 mL
FIS13-681-5272.5 to 30 mL
FBD136815285 to 60 mL
FIS13-681-5285 to 60 mL
Catalog number FBD13681529
Price (MYR)
1,928.00
Add to cart
Volume (Metric):
10 to 100 mL
Price (MYR)
1,928.00
Add to cart
  • Re-circulation valve helps user in quick purging without wasting any reagent
  • Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and minimizing the maintainence
  • Presence of unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, ensures wiper like smooth and effortless piston movement
  • Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
  • Adjustable delivery nozzle provides flexibility to users in demanding laboratory conditions
  • Bottle neck adapters of different sizes (28, 30, 32, 38, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
  • Each dispenser is accompanied with tool, which helps in easy and quick re-calibration
  • Contemporary and well designed contours, silky surface finish
  • Individually calibrated as per ISO 8655-2
  • Autoclavable 
Storage:
  • Store instrument and accessories only in clean conditions in cool and dry place (from -20°C to 50°C)
Specifications
Product TypeBottletop Dispenser
AutoclavableAutoclavable
Certifications/ComplianceIndividually calibrated as per ISO 8655-2.
ColorWhite and Fisher Blue
Density2.2 g/cm3
DescriptionContemporary and well designed contours. Silky surface finish
MaterialPolypropylene - Outer Body (10% Glass Filled)
Volume (Metric)10 to 100 mL
For Use With (Equipment)Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel.
IncludesCalibration tool, Adapters set (5 nos.), Instruction Manual, Inlet tube, Calibration report
ShapeCylindrical
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Vapor Pressure300mbar to 600mbar
Viscosity Rating500mm2
Warranty1 year free replacement warranty against any manufacturing defects.
Unit Size