Bottle Top Dispenser with Re-Circulation Valve
Bottle Top Dispenser with Re-Circulation Valve
Actual product may vary
Fisherbrand
Bottle Top Dispenser with Re-Circulation Valve
Provides a contemporary design coupled with a recirculation valve. Bottle Top Dispenser with Re-Circulation Valve has enhanced chemical compatibility and smooth piston movement.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FBD136815285 to 60 mL
FIS13-681-5250.5 to 5 mL
FBD136815261 to 10 mL
FIS13-681-5261 to 10 mL
FIS13-681-5272.5 to 30 mL
FIS13-681-5285 to 60 mL
FBD1368152910 to 100 mL
Catalog number FBD13681528
Price (MYR)
1,365.00
Add to cart
Volume (Metric):
5 to 60 mL
Price (MYR)
1,365.00
Add to cart
  • Re-circulation valve helps user in quick purging without wasting any reagent
  • Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and minimizing the maintainence
  • Presence of unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, ensures wiper like smooth and effortless piston movement
  • Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
  • Adjustable delivery nozzle provides flexibility to users in demanding laboratory conditions
  • Bottle neck adapters of different sizes (28, 30, 32, 38, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
  • Each dispenser is accompanied with tool, which helps in easy and quick re-calibration
  • Contemporary and well designed contours, silky surface finish
  • Individually calibrated as per ISO 8655-2
  • Autoclavable 
Storage:
  • Store instrument and accessories only in clean conditions in cool and dry place (from -20°C to 50°C)
Specifications
Product TypeBottletop Dispenser
AutoclavableAutoclavable
Certifications/ComplianceCE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025
ColorWhite and Fisher Blue
Density2.2 g/cm3
DescriptionShape: Cylindrical
MaterialPolypropylene - Outer Body (10% Glass Filled)
Volume (Metric)5 to 60 mL
For Use With (Equipment)Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel.
IncludesCalibration tool, Adapters set (5 nos.), Instruction Manual, Inlet tube, Calibration report
Vapor Pressure300mbar to 600mbar
Viscosity Rating500mm2
Warranty1 year free replacement warranty against any manufacturing defects.
Unit Size