Autoclavable Bottletop Dispensers
Autoclavable Bottletop Dispensers
Actual product may vary
Fisherbrand
Autoclavable Bottletop Dispensers
Provides smooth, effortless dispensing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAccuracyIncrementsPrecisionVolume (Metric)
FBD1361512±0.6% (±0.060mL)0.2 mL±0.2% (±0.020mL)1 to 10 mL
FBD1361510±0.6% (±0.015mL)0.05 mL±0.2% (±0.005mL)0.25 to 2.5 mL
FIS13-615-10±0.6% (±0.015mL)0.05 mL±0.2% (±0.005mL)0.25 to 2.5 mL
FBD1361511±0.6% (±0.030mL)0.1 mL±0.2% (±0.010mL)0.5 to 5 mL
FIS13-615-11±0.6% (±0.030mL)0.1 mL±0.2% (±0.010mL)0.5 to 5 mL
FBD1361513±0.6% (±0.180mL)0.5 mL±0.2% (±0.060mL)2.5 to 30 mL
FBD1361514±0.6% (±0.360mL)1.0 mL±0.2% (±0.120mL)5 to 60 mL
FBD1361515±0.6% (±3.000mL)2.0 mL±0.2% (±0.200mL)10 to 100 mL
Catalog number FBD1361512
Price (MYR)
1,023.00
Add to cart
Accuracy:
±0.6% (±0.060mL)
Increments:
0.2 mL
Precision:
±0.2% (±0.020mL)
Volume (Metric):
1 to 10 mL
Price (MYR)
1,023.00
Add to cart
Provides smooth, effortless dispensing
  • Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
  • 7 volume ranges to choose from
  • Bubble-free dispensing
  • Effortless plunger movement
  • Disassembles for easy cleaning and servicing
  • Autoclavable at 121°C, 15 psi for 10 to 15 minutes
  • Each bottle top dispenser includes one 28mm, 32mm, 38mm, 40mm, 45mm bottle neck adaptor
Component Description
  • Piston: PTFE
  • Cylinder: Glass
  • Volume Adjustment Knob: Screw Type
  • Valve Housing: PTFE
  • Valve Assembly: Glass Ball & Seal
  • Discharge Assembly: PTFE
  • Delivery Tube: FEP
  • Inlet Tube: FEP, 13 in. (with extension) 
  • Calibration: Individually calibrated and certified (in-lab by user is also possible)
  • Accuracy & Reproducibility: In accordance with ISO 8655 standards
  • Compability: Excellent with most reagents
Specifications
Accuracy±0.6% (±0.060mL)
Product TypeAnalog Bottletop Dispenser
AutoclavableAutoclavable
Channels1
MaterialPTFE, FEP, Borosilicate Glass
Volume (Metric)1 to 10 mL
Includes13-in. inlet tube (with extension)
Increments0.2 mL
Precision±0.2% (±0.020mL)
Unit Size