Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) | Culture Area |
---|---|---|
FB012925 | 150 mm | 143 sq. cm |
LSVFB012925 | 150 mm | 143 sq. cm |
LSVFB012920 | 35 mm | 8.5 sq. cm |
FB012921 | 60 mm | 21.2 sq. cm |
LSVFB012921 | 60 mm | 21.2 sq. cm |
FB012924 | 100 mm | 60.8 sq. cm |
LSVFB012924 | 100 mm | 60.8 sq. cm |