Thermolyne™ Largest Tabletop Muffle Furnaces
Thermolyne™ Largest Tabletop Muffle Furnaces
Actual product may vary
Thermo Fisher Scientific
Thermolyne™ Largest Tabletop Muffle Furnaces
Triple the usable working area with two supplied accessory refractory shelves using Thermolyne Largest Tabletop Muffle Furnace. Available in three temperature control models.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature ControlVoltageCertifications/Compliance
F30430CMDigital programmable w/4 programs, 16 segments each (D1)240 VCSA approved
F30420CDigital single setpoint w/ramp and dwell (B1)240 VCSA approved
F30420C-33Digital single setpoint w/ramp and dwell (B1)240 VCSA approved, CE marked
F30420C-33-80Digital programmable w/1 program, 8 segments (C1)240 VCSA approved, CE marked
F30420C-80Digital programmable w/1 program, 8 segments (C1)240 VCSA approved
F30430CM-33Digital programmable w/4 programs, 16 segments each (D1)240 VCSA approved, CE marked
Catalog number F30430CM
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Control:
Digital programmable w/4 programs, 16 segments each (D1)
Voltage:
240 V
Certifications/Compliance:
CSA approved
Triple your usable working area with two supplied accessory refractory shelves using Thermo Scientific™ Thermolyne™ Largest Tabletop Muffle Furnace. Designed for safe operation, these furnaces are available in three temperature control models. They are ideal for annealing glass, determinations of volatiles, catalyst research and ashing organic and inorganic samples.
Key Features
  • Advanced LED digital-set/digital-display temperature controller is microprocessor-controlled
  • LED display simultaneously shows both setpoint and actual furnace temperatures in °C or °F
  • User-selectable overtemperature protection
  • Open thermocouple protection
  • Adjustable power output from 1 to 100%
Safety and Design Features
  • Heating elements are on chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of chamber incorporates a 0.95cm (0.38in.) diameter port for monitoring chamber temperatures with independent measuring devices
  • Critical electronic components and heating elements are protected by a 35A circuit breaker
  • Door safety switch stops power to the heating elements when door opens
Choice of Temperature Controllers
  • Furnaces that use B1, C1 or D1 control also use a mechanical Overtemperature Protection relay
  • B1: Digital single setpoint control with a single ramp to setpoint and dwell; single display shows actual temperature or setpoint
  • C1: Digital programmable control with one stored program of 8 segments
  • D1: Digital programmable control with 4 stored programs, 16 segments per program, and RS-232 communications interface
Includes:
Two refractory shelves
Requires:
Electrical connection requires fixed wiring (no power cord).
Recommended for:
Annealing glass, determinations of volatiles, catalyst research and ashing organic and inorganic samples.
Specifications
Capacity (English)1.6 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF30430CM
Capacity (Metric)45 L
Temperature (Metric)100°C to 1093°C
TypeMultisegment, Multiprogram
Power Consumption550 w
Temperature ControlDigital programmable w/4 programs, 16 segments each (D1)
Temperature Stability±1.2 at 1000°C
Temperature Uniformity±3.45°C
Dimensions (D x W x H) Interior14 x 14 x 14 in. (36 x 36 x 36 cm)
Dimensions (L x W x H) Exterior25.5 x 21.5 x 29.5 in. (64.7 x 54.6 x 74.9 cm)
Length (English)25.5 in.
Length (Metric)64.7 cm
Width (English)14 in.
Width (Metric)54.6
Width (Metric) Interior36 cm
Height (English)29.5 in.
Height (English) Interior14 in.
Height (Metric)74.9 cm
Height (Metric) Interior36 cm
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Shipping Weight (English)260 lb.
Shipping Weight (Metric)117.9 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage22.9 A