2mL Nonsterile Centrifugal Filters
2mL Nonsterile Centrifugal Filters
Actual product may vary
2mL Nonsterile Centrifugal Filters
Filter volumes up to 2mL with these 2mL polypropylene centrifugal filters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Pore Size
F2520-3
F2520-1Cellulose Acetate0.22 μm
NSCF2520-1
F2520-2
NSCF2520-2
NSCF2520-3
Catalog number F2520-3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Filter volumes up to 2mL with these Thermo Scientific™ National 2mL Nonsterile Centrifugal Filters. The filters feature a virgin polypropylene filter housing with tapered 5mL, capped receiver tube, and can be used with any benchtop or floor model laboratory centrifuge.
  • 5,000xG maximum centrifugal force
  • Nonsterile
  • Bulk packed
Specifications
Volume (Metric)2 mL
Unit Size