2mL Nonsterile Centrifugal Filters
2mL Nonsterile Centrifugal Filters
Actual product may vary
2mL Nonsterile Centrifugal Filters
Filter volumes up to 2mL with these 2mL polypropylene centrifugal filters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Pore Size
F2520-1Cellulose Acetate0.22 μm
F2520-3Nylon0.2 μm
NSCF2520-1Cellulose Acetate0.22 μm
F2520-2Cellulose Acetate0.45 μm
NSCF2520-2Cellulose Acetate0.45 μm
NSCF2520-3Nylon0.2 μm
Catalog number F2520-1
Price (MYR)
-
Material (Membrane):
Cellulose Acetate
Pore Size:
0.22 μm
Filter volumes up to 2mL with these Thermo Scientific™ National 2mL Nonsterile Centrifugal Filters. The filters feature a virgin polypropylene filter housing with tapered 5mL, capped receiver tube, and can be used with any benchtop or floor model laboratory centrifuge.
  • 5,000xG maximum centrifugal force
  • Nonsterile
  • Bulk packed
Specifications
DescriptionCellulose Acetate Membrane; 0.22µm Pore Size
For Use With (Equipment)Benchtop or Floor Model Centrifuge
Material (Membrane)Cellulose Acetate
Pore Size0.22 μm
SterilityNon-sterile
Housing MaterialPolypropylene
Product LineTarget2
TypeCentrifugal Filter
Volume (Metric)2 mL
Unit Size