EcoPolyBlend™ Collection Center
EcoPolyBlend™ Collection Center
Actual product may vary
EcoPolyBlend™ Collection Center
EcoPolyBlend™ Collection Center
EcoPolyBlend™ Collection Center
All-weather construction stands up to the harshest environments
Have Questions?
Catalog NumberCapacity (English)Holds
ERW2868566 gal.55 gal. Drum
Catalog number ERW28685
Price (MYR)
1,474.00
EA
Add to cart
Capacity (English):
66 gal.
Holds:
55 gal. Drum
Price (MYR)
1,474.00
EA
Add to cart
  • Large opening for easy loading and empty bases nest for storage
  • 100% recycled polyethylene
  • Color: Black
Single Drum Collection
  • Accommodates one 208.2L (55 gal.) drum
  • Sump Capacity: 250L (66 gal.)
  • Dimensions (Dia. x H): 83.8 x 80.6cm (33 x 31.75 in.)
Double Drum Collection
  • Accommodates two 208.2L (55 gal.) drums
  • Dual covers can be opened separately or closed together and locked
  • Forklift channels for relocating
  • Sump Capacity: 681L (180 gal.)
  • Dimensions (L x W x H): 152.4 x 99.1 x 119.4 cm (60 x 39 x 47 in.)
Specifications
Certifications/ComplianceSPCC, EPA, OSHA, UL
ColorBlack
Capacity (English)66 gal.
Capacity (Metric)250 L
MaterialPolyethylene
TypeSingle Drum Collection Center
For Use With (Application)Easy loading and empty bases nest for storage
Holds55 gal. Drum
Width (English)31 in.
Width (Metric)78.74 cm
Height (English)31.75 in.
Height (Metric)80.6 cm
Diameter (English)33 in.
Diameter (Metric)83.8 cm
Unit SizeEA