EZ-Link™ Biocytin
EZ-Link™ Biocytin
Actual product may vary
Erweka
EZ-Link™ Biocytin
Synthesize long or trifunctional biotinylation reagents using this biotin variant containing a primary amine in its valeric acid chain.
Have Questions?
Catalog NumberQuantity
ERW28022
Catalog number ERW28022
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Thermo Scientific™ EZ-Link Biocytin is a simple variant of biotin that contains a primary amine in its valeric acid chain, which provides the essential backbone for construction of certain long-chain and trifunctional biotinylation reagents.
Highlights:
  • Useful for synthesizing long-chain biotinylation reagents (can use EDC, Part No. 22980, 22981)
  • An extended spacer arm intermediate for further derivatization into reactive group-specific biotinylating reagents
  • Contains both carboxyl group and primary amine, which may be modified to make trifunctional reagents like Sulfo-SBED
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeBiotin & Analogs
Unit Size
Contents & Storage
Store at room temperature.