Immunomagnetic depletion of apoptotic (Annexin V+) cells from culture or tissue preparations
Have Questions?
Catalog Number
Quantity
ERW17899
1 x 109 Cells
Catalog number ERW17899
Price (MYR)
5,539.00
EA
Add to cart
Quantity:
1 x 109 Cells
Price (MYR)
5,539.00
EA
Add to cart
The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection
Unwanted cells are labeled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
Labeled cells are separated using an EasySep™ magnet without the use of columns
Desired cells are simply poured off into a new tube
Annexin V binds phosphatidylserine on the outer leaflet of the cell membrane during apoptosis