EasySep™ HLA Chimerism BC CD56 Positive Selection Kit
EasySep™ HLA Chimerism BC CD56 Positive Selection Kit
Actual product may vary
EasySep™ HLA Chimerism BC CD56 Positive Selection Kit
Column-free immunomagnetic positive selection of human CD56+ cells for HLA analysis
Have Questions?
Catalog NumberQuantity
ERW1787530 mL buffy coat
Catalog number ERW17875
Price (MYR)
2,780.00
EA
Add to cart
Quantity:
30 mL buffy coat
Price (MYR)
2,780.00
EA
Add to cart
  • The EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit II is designed to isolate CD56+ cells from freshly prepared buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD56 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD56 antigen is expressed on NK cells and NKT cells
  • This product replaces the EasySep™ Human Buffy Coat CD56 Positive Selection Kit (Catalog #18085) and EasySep™ HLA Buffy Coat CD56 Positive Selection Kit (Catalog #18085HLA), for even faster cell isolations
Specifications
Cell TypeNK Cells
Quantity30 mL buffy coat
Unit SizeEA