Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
Actual product may vary
Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
Nalgene FLASK FILTERING PP;1000ML ]] 1/CS
FLASK FILTERING PP;1000ML ]]
Have Questions?
Catalog number DS4101-1000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Nalgene FLASK FILTERING PP;1000ML ]]
  • Limited Quantity Transport Exemption No
  • Whether it is included in Item 2828 No
  • Whether it is a dangerous good NA
  • Whether it is likely to cause poisoning NA
  • Whether it is highly toxic NA
  • Whether it is likely to explode NA
Specifications
Capacity (English)32 oz.
DescriptionVacuum Flask, polypropylene, 1000mL; Flask Filtering PP 1000mL
Capacity (Metric)1000 mL
MaterialPolypropylene
No. per Case1
No. per Pack1
AutoclavableAutoclavable
Closure SizeStopper No. 8
Unit SizeEACH