Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Actual product may vary
Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure
Utilize these wide-mouth, amber HDPE economy bottles for light-duty, general-purpose applications where a heavier bottle is not required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)dimClosureSize
DS2185-0016500 mL
DS2185-00321 L
Catalog number DS2185-0016
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Conduct light-duty, general-purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Wide-Mouth Amber HDPE Economy Bottles with Closure. These high-density, polyethylene bottles are lightweight versions of the original Nalgene Amber Lab Quality bottle. Opaque amber color reduces UV light transmissions to protect light-sensitive liquids. Useful for storage, shipping and packaging, they offer excellent chemical resistance to most acids, bases and alcohols.
Features:
  • Economical and recyclable; designed for single use
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
  • Meets U.S. Pharmacopoeia 23 requirements for maximum light transmission
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
Amber polypropylene screw closures
.
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypeScrew Cap
Diameter (Metric)73 mm
Height (English)6.62 in.
Height (Metric)168 mm
Through the Closure DispensingNo
Width (English)2.87 in.
Product TypeWide Mouth Economy Bottle
Capacity16 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorAmber
DescriptionAmber Wide-Mouth Economy Bottle
Diameter (Metric) Inner Neck44 mm
MaterialHDPE
No. per Case48
No. per Pack48
SterilityNon-sterile
Closure ColorAmber
Neck Finish53mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2003 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use. USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableSingle-use
GraduatedNo
MouthWide
ShapeRound
Unit Size48