Barnstead™ Hose Nipple Cartridges
Barnstead™ Hose Nipple Cartridges
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Hose Nipple Cartridges
Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionResin Type
D8901Full Size High CapacityTwo Bed Ion Exchange
D8902Full Size UltrapureUltrapure Mixed Bed Ion Exchange
Catalog number D8901
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Full Size High Capacity
Resin Type:
Two Bed Ion Exchange
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8 in. barb hose connections on each end
  • D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
Specifications
DescriptionFull Size High Capacity
For Use With (Equipment)Barnstead Water Purification System
Particle Retention1650 grains
TypeCartridge, Hose Nipple
Flow RateUp to 75 L/hr.
For Use With (Application)Two Bed Ion Exchange
IncludesColor Indicator
Resin TypeTwo Bed Ion Exchange
Unit Size