Barnstead™ Nanopure™ Accessories and Consumables
Barnstead™ Nanopure™ Accessories and Consumables
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
D50280R/O and distilled feed; Ultralow organics, type I water
D50251Pretreatment Cartridge, tap and deionized water
D50253Ultrapure Deionization Cartridge
D50256Pretreatment Cartridge Kit, tap and deionized water
D50281Deionized water feed ; Ultralow organics, type I water
D50282R/O and distilled feed; Low organics, type I water
D50283Deionized water feed; Low organics, type I water
Catalog number D50280
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
R/O and distilled feed; Ultralow organics, type I water
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
DescriptionR/O and distilled feed; Ultralow organics, type I water
For Use With (Equipment)Nanopure Water Purifications System
TypeR/O and Distilled Feed
Unit Size