Polycarbonate Erlenmeyer Flasks
Polycarbonate Erlenmeyer Flasks
Actual product may vary
Corning
Polycarbonate Erlenmeyer Flasks
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Outer NeckClosure TypeGraduations
CRG430421125 mL2.6 cmFlat25 mL
CRG430183250 mL3.1 cmFlat25 mL
CRG430422500 mL4.3 cmFlat50 mL
Catalog number CRG430421
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
125 mL
Diameter (Metric) Outer Neck:
2.6 cm
Closure Type:
Flat
Graduations:
25 mL
  • Made from optically clear polycarbonate
  • Ideal for shaker culture applications
  • Available with flat caps or vented caps for applications requiring sterile gas exchange
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase- and DNase-free
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)4.22 oz.
Closure ColorOrange
Closure MaterialPolypropylene
DescriptionTop style: Threaded
Diameter (Metric) Outer Neck2.6 cm
Capacity (Metric)125 mL
MaterialPolycarbonate
For Use With (Application)For shaker culture applications
Graduations25 mL
Neck StyleWide Mouth
No. per Case50/Cs.
No. per Pack1/Pk.
DisposableSingle-use
Mouth StyleWide
SterilitySterile
Closure TypeFlat
Unit SizeCS