Disposable Polystyrene Serological Pipets, Sterile
Disposable Polystyrene Serological Pipets, Sterile
Actual product may vary
Corning
Disposable Polystyrene Serological Pipets, Sterile
Features exceptionally crisp, dark, easy-to-read printing with reverse and negative graduations. Falcon™ Disposable Polystyrene Serological Pipets are individually wrapped.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Graduations
CRG3575211 mL0.01 mL
CRG35755110 mL0.1 mL
CRG35752525 mL0.25 mL
Catalog number CRG357521
Price (MYR)
1,463.10
Special offer
Online exclusive
Ends: 31-Aug-2025
2,238.00
Save 774.90 (35%)
CS
Add to cart
Volume (Metric):
1 mL
Graduations:
0.01 mL
Price (MYR)
1,463.10
Special offer
Online exclusive
Ends: 31-Aug-2025
2,238.00
Save 774.90 (35%)
CS
Add to cart
  • Polystyrene, disposable pipets for tissue culture, bacteriological, and research applications
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0.1 EU/mL
  • BSE/TSE-free
  • Negative graduations for extra capacity
  • Reverse graduations (except 1 mL)
  • Color-coded package and markings for ease of identification
  • Polyester plug to help prevent overfill
  • Accurate ±2% at full volume (excluding 1 and 2 mL)
Graduation alignment system
The pipet graduation alignment system is a fast and simple way to bring graduations to your line of sight. Simply pop open or peel back the top of the pipet package and line up the pipettor before completely removing the pipet from its wrapping. When you remove the pipet from its package, the graduations should face you every time.
Specifications
Accuracy±3%
Aspirator PlugYes
Certifications/ComplianceNon-pyrogenic, ISO 9001: 2000, FDA, 820 CFR
Coefficient of Variation1.5%
MaterialPolystyrene
SterilitySterile
For Use With (Application)For tissue culture usage or any non toxic solution
GraduatedYes
Graduations0.01 mL
Negative Graduations0.4 mL
Quantity1000/Cs.
Tip StyleStandard
Tolerance±2%
Packaging TypeIndividually Wrapped, Paper-Plastic
ColorYellow
Volume (Metric)1 mL
TypeSerological Pipet