WRP-50 All-Hazards Alert (NOAA) Radio
WRP-50 All-Hazards Alert (NOAA) Radio
Actual product may vary
WRP-50 All-Hazards Alert (NOAA) Radio
Turns on automatically to supply instant alerts to dangerous situations.
Have Questions?
Catalog number BSN70900
Price (MYR)
1,403.00
EA
Add to cart
Price (MYR)
1,403.00
EA
Add to cart
  • Broadcasts warnings plus information from the government on disaster precautions and procedures
  • Specific Area Message Encoding (S.A.M.E.) eliminates false alerts
  • Recommended by the U.S. government for all public/private facilities
  • Provides 24-hour weather updates at the touch of a button
  • Mounts on desk or wall
  • 120V with 9V battery backup
  • Dimensions: 11.5L x 6.5W x 4 in.H (38.1 x 16.5 x 10.2cm)
Specifications
Height (English)4 in.
Height (Metric)10.2 cm
Voltage120 V
Width (English)6.5 in.
Width (Metric)16.5 cm
Unit SizeEA