Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Actual product may vary
Thermo Fisher Scientific
Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
B2-24Double-sided Comb241.0/1.5 mm
B2-20Double-sided Comb201.0/1.5 mm
B2-RL-25DRapid Load Comb25 (2X)1.5 mm
Catalog number B2-24
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Double-sided Comb
Teeth Count:
24
Thickness (Metric) Teeth:
1.0/1.5 mm
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Ordering Information:
  • Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8 and 12-channel pipette format (1X) and (2X) combs
Specifications
For Use With (Equipment)Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Gel SizeMini
Product LineOwl, EasyCast
TypeDouble-sided Comb
Teeth Count24
Thickness (Metric) Teeth1.0/1.5 mm
Unit Size