Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Actual product may vary
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Thermo Fisher Scientific
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with these B1A Mini Gel Electrophoresis systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
B1AEasyCast
B1A-BPEasyCast w/ Buffer Exchange Ports
Catalog number B1A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Includes:
EasyCast
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems. The ideal size for every lab, this system determines molecular weight ranges of PCR fragments or larger sized DNA molecules.
Features:
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
  • Leakproof casting allows a gel to be cast and run in the same chamber with no tape or additional parts required
  • Run from five to 24 samples in 15 minutes.
Ordering Information:
Available with optional buffer exchange ports (B1A-BP) for use with an external recirculation pump.
Includes:
  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast gasketed UVT gel tray
  • Two heavy-duty, one-piece, double-sided combs: 6 & 10 well, 1.0/1.5mm thick
Specifications
Certifications/ComplianceCE marked
Gel SizeMini
Product LineOwl
TypeMini Gel Electrophoresis System
IncludesEasyCast
Sample Size5 to 24
Volume (Metric) Buffer400 mL
Dimensions (L x W x H)6.31 x 4.13 x 3.75 in. (16 x 10.5 x 9.5 cm)
Length (English)6.31 in.
Length (Metric)16 cm
Length (Metric) Gel8 cm
Width (English)4.13 in.
Width (Metric)10.5 cm
Width (Metric) Gel7 cm
Height (English)3.75 in.
Height (Metric)9.5 cm
Max. Voltage150 V
Unit Size