Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Actual product may vary
Thermo Fisher Scientific
Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty one-piece combs, for use with the Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
B1-12CStandard Comb121 mm
B1-10Double-sided Comb101.0/1.5 mm
B1-12DStandard Comb121.5 mm
B1-5CStandard Comb51 mm
B1-5DStandard Comb51.5 mm
B1-8CStandard Comb81 mm
B1-8DStandard Comb81.5 mm
Catalog number B1-12C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Standard Comb
Teeth Count:
12
Thickness (Metric) Teeth:
1 mm
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
Ordering Information:
  • Cat Nos. B1-Rl-9C, -9D, -17C and -17D are 8 and 12-channel pipette format (1X) and (2X) combs
  • Customized combs are available (Order SKU # XCM); please contact your customer service or sales representative for further details
Specifications
DescriptionStandard 12 tooth comb; 1.0mm thick
For Use With (Equipment)Owl™ EasyCast B1 Mini Gel Electrophoresis System
Gel SizeMini
Product LineOwl, EasyCast
TypeStandard Comb
Teeth Count12
Thickness (Metric) Teeth1 mm
Unit Size