Search Thermo Fisher Scientific
Catalog Number | Type | Teeth Count | Thickness (Metric) Teeth |
---|---|---|---|
B1-10 | Double-sided Comb | 10 | 1.0/1.5 mm |
B1-12C | Standard Comb | 12 | 1 mm |
B1-12D | Standard Comb | 12 | 1.5 mm |
B1-5C | Standard Comb | 5 | 1 mm |
B1-5D | Standard Comb | 5 | 1.5 mm |
B1-8C | Standard Comb | 8 | 1 mm |
B1-8D | Standard Comb | 8 | 1.5 mm |
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
Ordering Information: