Check Order Status
Tetrahydrofuran (HPLC), Fisher Chemical™
Element Search
Structure Search
Alfa Aesar
Tetrahydrofuran (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change view
Quantity:
4 L
1 L
Packaging:
View all
Glass Bottle
Catalog number
ALFT425-4
Price (MYR)
243.10
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
243.10
Add to cart
Specifications
Chemical Name or Material
Tetrahydrofuran
pH
7 to 8
Cleanup Symbol
Solvent
Melting Point
-108.4°C
Absorbance
Pass Test, 1.0 AU max. at 210nm, 0.6 AU max. at 215nm, 0.3 AU max. at 230nm, 0.1 AU max. at 254nm
View more