4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
CAS: 16208-17-6 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL20274.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS632-99-5
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC20H20ClN3
InChI KeyAXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial sourced in the U.S. and in other countries
lambda-max (UV-Vis)547-557 nm
Appearance (Color)Dark green
Specific Absorbance (E 1%/1cm)≥1500 at lambda max (50% aq. ethanol, non-U.S. sourced material)
FormCrystals or powder or crystalline powder